Learn More
Benchmark Products Inc Benchmark Suite Pad Mop, Validated Sterile, Effective Cleanroom Use, 15 in
SDP
Catalog No. NC1411449
Encompass
The Benchmark Products Inc. Suite Pad Mop is a validated sterile mop designed for critical applications in cleanrooms and controlled environments. Engineered with unique synthetic ultra-fibers, it offers over 98% contamination removal proficiently while minimizing cross-contamination to less than 0.02%. Its snag-resistant and absorbent material ensures superior cleaning efficacy, making it ideal for disinfectant application across various surfaces like walls, ceilings, and floors.
Catalog No. NC1411449 Supplier Benchmark Products Inc Supplier No. YGLPP51525S
The Benchmark Products Inc. Suite Pad Mop is a validated sterile mop designed for critical applications in cleanrooms and controlled environments. Engineered with unique synthetic ultra-fibers, it offers over 98% contamination removal proficiently while minimizing cross-contamination to less than 0.02%. Its snag-resistant and absorbent material ensures superior cleaning efficacy, making it ideal for disinfectant application across various surfaces like walls, ceilings, and floors.