Learn More
GENEPATH DIAGNOSTICS INC Sul1, Sul2 quantitative standards for assay calibration and validation. Provided at high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL) concentrations (RUO)
Catalog No. NC3949389
Encompass_Preferred
Synthetic, sequence-verified quantitative standards targeting the sul1, sul2 gene associated with sulfonamide resistance. Supplied at defined concentrations of High – 10,000 copies/µL, Medium – 1,000 copies/µL, and Low – 100 copies/µL, these standards are intended for accurate assay calibration and quantitative performance evaluation across a broad dynamic range. They enable robust assay validation, sensitivity and limit-of-detection (LoD) determination, and assessment of assay linearity in molecular assays. Compatible with real-time PCR and other nucleic acid amplification platforms, the standards ensure consistent inter-run comparability, reliable quantification, and ongoing performance monitoring for research, assay development, and routine quality control applications in antimicrobial resistance testing (RUO)
Catalog No. NC3949389 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10890080050
Synthetic, sequence-verified quantitative standards targeting the sul1, sul2 gene associated with sulfonamide resistance. Supplied at defined concentrations of High – 10,000 copies/µL, Medium – 1,000 copies/µL, and Low – 100 copies/µL, these standards are intended for accurate assay calibration and quantitative performance evaluation across a broad dynamic range. They enable robust assay validation, sensitivity and limit-of-detection (LoD) determination, and assessment of assay linearity in molecular assays. Compatible with real-time PCR and other nucleic acid amplification platforms, the standards ensure consistent inter-run comparability, reliable quantification, and ongoing performance monitoring for research, assay development, and routine quality control applications in antimicrobial resistance testing (RUO)