Learn More
U.S. Pharmacopeia Sulbactam, 68373-14-8, MFCD00867005, 250mg
Catalog No. NC9041272
Encompass
Molecular formula C8H11NO5S, Molecular weight 233.24, Melting Point 298.4 - 303.8 °F (148 - 151 °C) (also reported as 170 °C), Synonyms: Penicillanic acid 1,1-dioxide. Cold Shipment Required
Catalog No. NC9041272 Supplier U.S. Pharmacopeia Supplier No. 1623670
Molecular formula C8H11NO5S, Molecular weight 233.24, Melting Point 298.4 - 303.8 °F (148 - 151 °C) (also reported as 170 °C), Synonyms: Penicillanic acid 1,1-dioxide. Cold Shipment Required