Learn More
Selleck Chemical LLC Sulbactam S1958-10mg
SDP
Supplier:  Selleck Chemical LLC S195810MG
Encompass_Preferred
Sulbactam(CP45899) is a beta-lactamase inhibitor with an average IC50 of 0 8 M
Catalog No. 50-310-8584