Learn More
Selleck Chemical LLC Sulbactam S1958-200mg
SDP
Catalog No. 503110925
Encompass_Preferred
Sulbactam(CP45899) is a beta-lactamase inhibitor with an average IC50 of 0 8 M
Catalog No. 50-311-0925 Supplier Selleck Chemical LLC Supplier No. S1958200MG
Sulbactam(CP45899) is a beta-lactamase inhibitor with an average IC50 of 0 8 M