Learn More
Medchemexpress LLC Sulbactam | 68373-14-8 | 100.0% | 1 ML
SDP
Catalog No. 5000406710
Encompass_Preferred
Sulbactam is a competitive, irreversible beta-lactamase inhibitor that demonstrates antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex. It exhibits a wide range of antibacterial activity against both Gram-positive and Gram-negative aerobic and anaerobic bacteria.
  • Competitive, irreversible beta-lactamase inhibitor
  • Antimicrobial against multidrug-resistant Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex
  • Wide range of antibacterial activity
  • Active against Gram-positive and Gram-negative aerobic and anaerobic bacteria
  • For research use only
Catalog No. 50-004-06710 Supplier Medchemexpress LLC Supplier No. HYB033410MM/1ML
Sulbactam is a competitive, irreversible beta-lactamase inhibitor that demonstrates antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex. It exhibits a wide range of antibacterial activity against both Gram-positive and Gram-negative aerobic and anaerobic bacteria.
  • Competitive, irreversible beta-lactamase inhibitor
  • Antimicrobial against multidrug-resistant Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex
  • Wide range of antibacterial activity
  • Active against Gram-positive and Gram-negative aerobic and anaerobic bacteria
  • For research use only