Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulbactam | 68373-14-8 | 100.0% | 100 MG
SDP

Catalog No. 5000406598
Encompass_Preferred

Sulbactam is a competitive, irreversible beta-lactamase inhibitor. It exhibits antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex and possesses a broad spectrum of antibacterial activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. It is suitable for use as a laboratory chemical and in the manufacture of substances.

  • Competitive, irreversible beta-lactamase inhibitor
  • Antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex
  • Broad spectrum antibacterial activity
  • Solid form, light yellow to yellow in color
  • Suitable for laboratory chemical applications
  • Melting point: 154-157°C
  • Boiling point: 356°C at 760 mmHg
  • Flash point: 297.1°C
  • For research use only

Catalog No. 50-004-06598 Supplier Medchemexpress LLC Supplier No. HYB0334100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulbactam is a competitive, irreversible beta-lactamase inhibitor. It exhibits antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex and possesses a broad spectrum of antibacterial activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. It is suitable for use as a laboratory chemical and in the manufacture of substances.

  • Competitive, irreversible beta-lactamase inhibitor
  • Antimicrobial activity against multidrug-resistant (MDR) Acinetobacter calcoaceticus-Acinetobacter baumannii (Acb) complex
  • Broad spectrum antibacterial activity
  • Solid form, light yellow to yellow in color
  • Suitable for laboratory chemical applications
  • Melting point: 154-157°C
  • Boiling point: 356°C at 760 mmHg
  • Flash point: 297.1°C
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.