Learn More
Medchemexpress LLC Sulbactam-d5 sodium | 1322625-44-4 | 99.9% | 500 UG
SDP
Catalog No. 5000405513
Encompass_Preferred
Sulbactam-d5 sodium is a deuterium-labeled form of Sulbactam sodium, a competitive, irreversible beta-lactamase inhibitor. It demonstrates antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii complex. Deuteration in drug molecules can impact pharmacokinetic and metabolic profiles, making this compound valuable for research.
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Deuterium stable isotope-labeled inhibitor for in vivo cell experiments
  • Applicable in therapeutic drug monitoring
Catalog No. 50-004-05513 Supplier Medchemexpress LLC Supplier No. HYB0334AS500UG
Sulbactam-d5 sodium is a deuterium-labeled form of Sulbactam sodium, a competitive, irreversible beta-lactamase inhibitor. It demonstrates antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii complex. Deuteration in drug molecules can impact pharmacokinetic and metabolic profiles, making this compound valuable for research.
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Deuterium stable isotope-labeled inhibitor for in vivo cell experiments
  • Applicable in therapeutic drug monitoring