Learn More
Medchemexpress LLC Sulbactam pivoxil | 69388-79-0 | MFCD00867003 | 98.0% | 347.38 | C14H21NO7S | 100 MG
SDP
Catalog No. 5000242350
Encompass_Preferred
Sulbactam pivoxil is the pivoxil ester prodrug of sulbactam, a β-lactamase inhibitor supplied as a research/analytical standard. The material is provided as a white to off-white solid with reported purity of 98.0% and a molecular weight of 347.38, and is used to support studies of oral bioavailability, pharmacokinetics, and β-lactamase inhibition.
  • Prodrug form improves oral absorption compared with sulbactam.
  • Useful as a reference standard for pharmacokinetic and microbiology studies.
  • High reported purity supports analytical and formulation work.
  • Available in solid and ready-to-use solution formats for flexibility.
  • Recommended storage conditions provided to preserve stability.
Catalog No. 50-002-42350 Supplier Medchemexpress LLC Supplier No. HY108288100MG
Sulbactam pivoxil is the pivoxil ester prodrug of sulbactam, a β-lactamase inhibitor supplied as a research/analytical standard. The material is provided as a white to off-white solid with reported purity of 98.0% and a molecular weight of 347.38, and is used to support studies of oral bioavailability, pharmacokinetics, and β-lactamase inhibition.
  • Prodrug form improves oral absorption compared with sulbactam.
  • Useful as a reference standard for pharmacokinetic and microbiology studies.
  • High reported purity supports analytical and formulation work.
  • Available in solid and ready-to-use solution formats for flexibility.
  • Recommended storage conditions provided to preserve stability.