Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulbactam pivoxil | 69388-79-0 | MFCD00867003 | 98.0% | 347.38 g·mol⁻¹ | C14H21NO7S | 1 ML
SDP

Catalog No. 5000242481
Encompass_Preferred

Sulbactam pivoxil is the pivoxil prodrug of sulbactam, a β-lactamase inhibitor used in antibacterial research. It is supplied for research and analytical applications and is available as an analytical solid or as a ready-to-use 10 mM solution in DMSO.

  • Available as solid and 10 mM solution in DMSO.
  • Purity 98.0%.
  • Molecular weight 347.38 g·mol⁻¹.
  • Chemical formula C14H21NO7S.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-42481 Supplier Medchemexpress LLC Supplier No. HY10828810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulbactam pivoxil is the pivoxil prodrug of sulbactam, a β-lactamase inhibitor used in antibacterial research. It is supplied for research and analytical applications and is available as an analytical solid or as a ready-to-use 10 mM solution in DMSO.

  • Available as solid and 10 mM solution in DMSO.
  • Purity 98.0%.
  • Molecular weight 347.38 g·mol⁻¹.
  • Chemical formula C14H21NO7S.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.