Learn More
Selleck Chemical LLC Sulbactam sodium S2551-50mg
SDP
Catalog No. 503111639
Encompass_Preferred
Sulbactam sodium (Unasyn CP-45899-2) is an irreversible -lactamase inhibitor
Catalog No. 50-311-1639 Supplier Selleck Chemical LLC Supplier No. S255150MG
Sulbactam sodium (Unasyn CP-45899-2) is an irreversible -lactamase inhibitor