Learn More
Medchemexpress LLC Sulbactam sodium | 69388-84-7 | 99.4% | 50 MG
SDP
Catalog No. 5000406766
Encompass_Preferred
Sulbactam sodium (Standard) is an analytical standard intended for research and analytical applications. It functions as a competitive, irreversible beta-lactamase inhibitor, demonstrating antimicrobial activity against multidrug-resistant (MDR) acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.
  • Used as a reference standard for assays
  • Competitive, irreversible beta-lactamase inhibitor
  • Shows antimicrobial activity against multidrug-resistant acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex
  • Utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-06766 Supplier Medchemexpress LLC Supplier No. HYB0334AR50MG
Sulbactam sodium (Standard) is an analytical standard intended for research and analytical applications. It functions as a competitive, irreversible beta-lactamase inhibitor, demonstrating antimicrobial activity against multidrug-resistant (MDR) acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.
  • Used as a reference standard for assays
  • Competitive, irreversible beta-lactamase inhibitor
  • Shows antimicrobial activity against multidrug-resistant acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex
  • Utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS