Learn More
Medchemexpress LLC Sulbactam sodium | 69388-84-7 | MFCD01750374 | 99.4% | 255.22 g/mol | C8H10NNaO5S | 10 MG
SDP
Catalog No. 5000298275
Encompass_Preferred
Sulbactam sodium is the sodium salt of sulbactam, supplied as a high-purity analytical standard for research and assay applications. It is characterized for use in method development, quality control, and analytical testing where a well-defined beta-lactamase inhibitor reference is required.
  • Sodium salt form of sulbactam.
  • Competitive, irreversible β-lactamase inhibitor.
  • Provided as an analytical reference standard for research and assays.
  • High assay purity (reported ~99.4%).
  • Available in small sizes suitable for laboratory use, e.g., 10 MG.
Catalog No. 50-002-98275 Supplier Medchemexpress LLC Supplier No. HYB0334AR10MG
Sulbactam sodium is the sodium salt of sulbactam, supplied as a high-purity analytical standard for research and assay applications. It is characterized for use in method development, quality control, and analytical testing where a well-defined beta-lactamase inhibitor reference is required.
  • Sodium salt form of sulbactam.
  • Competitive, irreversible β-lactamase inhibitor.
  • Provided as an analytical reference standard for research and assays.
  • High assay purity (reported ~99.4%).
  • Available in small sizes suitable for laboratory use, e.g., 10 MG.