Learn More
Medchemexpress LLC Sulbactam (sodium) (Standard) | 69388-84-7 | 99.4% | 25 MG
SDP
Catalog No. 5000450606
Encompass_Preferred
Sulbactam (sodium) (Standard) is an analytical standard intended for research and analytical applications. It acts as a competitive, irreversible beta-lactamase inhibitor, exhibiting antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.
  • Analytical standard for research and analytical applications
  • Competitive, irreversible beta-lactamase inhibitor
  • Exhibits antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS
Catalog No. 50-004-50606 Supplier Medchemexpress LLC Supplier No. 25MGHYB0334AR
Sulbactam (sodium) (Standard) is an analytical standard intended for research and analytical applications. It acts as a competitive, irreversible beta-lactamase inhibitor, exhibiting antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.
  • Analytical standard for research and analytical applications
  • Competitive, irreversible beta-lactamase inhibitor
  • Exhibits antimicrobial activity against multidrug-resistant Acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex
  • Used in qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS