Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulbenicillin disodium | 28002-18-8 | 25 MG
SDP

Catalog No. 5000420289
Encompass_Preferred

Sulbenicillin disodium is the disodium salt of Sulbenicillin, a Penicillin antibiotic. It exhibits antibacterial activity against various mucoid and non-mucoid strains of *Pseudomonas aeruginosa*. It has a molecular weight of 458.42 and appears as a white to off-white solid. This product is for research use only.

  • Exhibits antibacterial activity against *Pseudomonas aeruginosa*
  • Appearance: white to off-white solid
  • For research use only
  • Soluble in H2O at 250 mg/mL, may require sonication
  • Solid form storage: 4°C, sealed, away from moisture
  • Solution storage: -80°C for 6 months or -20°C for 1 month
  • Purity: 98.84%

Catalog No. 50-004-20289 Supplier Medchemexpress LLC Supplier No. 25MGHYN7097
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Sulbenicillin disodium is the disodium salt of Sulbenicillin, a Penicillin antibiotic. It exhibits antibacterial activity against various mucoid and non-mucoid strains of *Pseudomonas aeruginosa*. It has a molecular weight of 458.42 and appears as a white to off-white solid. This product is for research use only.

  • Exhibits antibacterial activity against *Pseudomonas aeruginosa*
  • Appearance: white to off-white solid
  • For research use only
  • Soluble in H2O at 250 mg/mL, may require sonication
  • Solid form storage: 4°C, sealed, away from moisture
  • Solution storage: -80°C for 6 months or -20°C for 1 month
  • Purity: 98.84%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.