Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulbenicillin disodium | 28002-18-8 | 50 MG
SDP

Catalog No. 5000418180
Encompass_Preferred

Sulbenicillin disodium is the disodium salt of Sulbenicillin. Sulbenicillin is a Penicillin antibiotic that exhibits antibacterial activity against various mucoid and non-mucoid strains of Pseudomonas aeruginosa.

  • Purity: 98.84%
  • Molecular Weight: 458.42
  • Formula: C16H16N2Na2O7S2
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
  • Solubility in H2O: 250 mg/mL (requires sonication)
  • Solubility in PBS: 100 mg/mL (clear solution; requires sonication)

Catalog No. 50-004-18180 Supplier Medchemexpress LLC Supplier No. HYN709750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulbenicillin disodium is the disodium salt of Sulbenicillin. Sulbenicillin is a Penicillin antibiotic that exhibits antibacterial activity against various mucoid and non-mucoid strains of Pseudomonas aeruginosa.

  • Purity: 98.84%
  • Molecular Weight: 458.42
  • Formula: C16H16N2Na2O7S2
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
  • Solubility in H2O: 250 mg/mL (requires sonication)
  • Solubility in PBS: 100 mg/mL (clear solution; requires sonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.