Learn More
Medchemexpress LLC Sulbenicillin disodium | 28002-18-8 | MFCD01682086 | 1 ML
SDP
Catalog No. 5000418105
Encompass_Preferred
Sulbenicillin disodium is the disodium salt of Sulbenicillin, a penicillin antibiotic. It is known for its antibacterial activity against various mucoid and non-mucoid strains of Pseudomonas aeruginosa. This product is typically supplied as a 10 mM solution in 1 mL of water, suitable for research and analytical applications.
  • Disodium salt of Sulbenicillin
  • Penicillin antibiotic
  • Exhibits antibacterial activity
  • Effective against Pseudomonas aeruginosa strains
  • Supplied as a pre-dissolved solution
Catalog No. 50-004-18105 Supplier Medchemexpress LLC Supplier No. HYN709710MM/1ML
Add to Cart
Add to Cart
Sulbenicillin disodium is the disodium salt of Sulbenicillin, a penicillin antibiotic. It is known for its antibacterial activity against various mucoid and non-mucoid strains of Pseudomonas aeruginosa. This product is typically supplied as a 10 mM solution in 1 mL of water, suitable for research and analytical applications.
  • Disodium salt of Sulbenicillin
  • Penicillin antibiotic
  • Exhibits antibacterial activity
  • Effective against Pseudomonas aeruginosa strains
  • Supplied as a pre-dissolved solution