Learn More
Medchemexpress LLC HY-B2229 1g Medchemexpress, Sulbutiamine CAS:3286-46-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB22291G
Encompass_Preferred
Medchemexpress, HY-B2229 1g Sulbutiamine CAS:3286-46-2 Sulbutiamine is a highly lipid soluble synthetic analogue of vitamin B1, used for the treatment of asthenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3118