Learn More
TARGETMOL CHEMICALS INC Sulcardine 2HCl 1MG
SDP
Catalog No. 502876616
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Sulcardine 2HCl is a multi-ion channel blocker with antiarrhythmic effects that blocks the properties of hERG and hNav1.5 channels and can be used to study atrial fibrillation and ventricular arrhythmias. Purity 99.51%
Catalog No. 50-287-6616 Supplier TARGETMOL CHEMICALS INC Supplier No. T839791MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Sulcardine 2HCl is a multi-ion channel blocker with antiarrhythmic effects that blocks the properties of hERG and hNav1.5 channels and can be used to study atrial fibrillation and ventricular arrhythmias. Purity 99.51%