Learn More
TARGETMOL CHEMICALS INC Sulcardine sulfate 25MG
SDP
Catalog No. 502879307
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sulcardine sulfate (B-87823) is a multi-ion channel blocker with antiarrhythmic activity, inhibits Na+, K+ and Ca2+ channels, and inhibits hNav1.5 channels in a concentration-dependent and reversible manner. Purity 98.74%
Catalog No. 50-287-9307 Supplier TARGETMOL CHEMICALS INC Supplier No. T2887925MG
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sulcardine sulfate (B-87823) is a multi-ion channel blocker with antiarrhythmic activity, inhibits Na+, K+ and Ca2+ channels, and inhibits hNav1.5 channels in a concentration-dependent and reversible manner. Purity 98.74%