Learn More
Medchemexpress LLC HY-B1460 100mg Medchemexpress, Sulconazole (mononitrate) CAS:61318-91-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1460100MG
Encompass_Preferred
Medchemexpress, HY-B1460 100mg Sulconazole (mononitrate) CAS:61318-91-0 Sulconazole mononitrate ((±)-Sulconazole mononitrate), an imidazole derivative, is a broad-spectrum fungicide and is inhibitory to certain gram-positive anaerobes. Sulconazole mononitrate can be used for the research of dermatomycoses, pityriasis versicolor, and cutaneous candidiasis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1195