Learn More
Medchemexpress LLC HY-107368 50mg Medchemexpress, Sulcotrione CAS:99105-77-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10736850MG
Encompass_Preferred
Medchemexpress, HY-107368 50mg Sulcotrione CAS:99105-77-8 Sulcotrione is a β-triketone herbicide which can inhibit hydroxyphenylpyruvate dioxygenase (HPPD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8390