Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfabenzamide, 100.1%, MP Biomedicals™

Catalog No. ICN10298125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10298125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10298125 Supplier MP Biomedicals, Inc Supplier No. 0210298125

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-71-9
Molecular Formula C13H12N2O3S
Molecular Weight (g/mol) 276.31
InChI Key PBCZLFBEBARBBI-UHFFFAOYSA-N
Synonym sulfabenzamide, sulfabenzide, sulfabenzid, sulfabenzoylamide, n-sulfanilylbenzamide, n-sulfamylbenzamide, n-benzoylsulfanilamide, sulfamylbenzamide, sulfabenzamid, sulfabenzamida
PubChem CID 5319
IUPAC Name N-(4-aminophenyl)sulfonylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Color White
Assay Percent Range 1.001
Quantity 25 g
Formula Weight 276.3
Physical Form Powder
Chemical Name or Material Sulfabenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.