Learn More
Medchemexpress LLC Sulfabenzamide-d4 (N-Sulfanilylbenzamide-d4) | 2732981-22-3 | 99.7% | 5 MG
SDP
Catalog No. 5000408292
Encompass_Preferred
Sulfabenzamide-d4 is a deuterium-labeled version of Sulfabenzamide, an antimicrobial agent effective against Gram-positive and Gram-negative bacterial strains. This compound is intended for research use only, serving as a tracer and internal standard in drug development and quantitative analysis. Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Antimicrobial agent
  • Effective against Gram-positive and Gram-negative bacterial strains
  • Used as a tracer in drug development
  • Internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08292 Supplier Medchemexpress LLC Supplier No. HYB0960S5MG
Sulfabenzamide-d4 is a deuterium-labeled version of Sulfabenzamide, an antimicrobial agent effective against Gram-positive and Gram-negative bacterial strains. This compound is intended for research use only, serving as a tracer and internal standard in drug development and quantitative analysis. Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Antimicrobial agent
  • Effective against Gram-positive and Gram-negative bacterial strains
  • Used as a tracer in drug development
  • Internal standard for quantitative analysis (e.g., NMR, GC-MS, LC-MS)
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs