Learn More
Medchemexpress LLC Sulfabrom-d4 | 2374858-36-1 | 361.25 | 10 MG
SDP
Catalog No. 5000448439
Encompass_Preferred
Sulfabrom-d4 is a deuterium labeled Sulfabrom, which acts as a long-acting Sulfonamide. It is employed for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.
  • Deuterium labeled compound
  • Long-acting Sulfonamide
  • Used for coccidiosis treatment
  • Used for bacterial infections in poultry, swine, and cattle
  • Store as powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-48439 Supplier Medchemexpress LLC Supplier No. HYU00131S10MG
Sulfabrom-d4 is a deuterium labeled Sulfabrom, which acts as a long-acting Sulfonamide. It is employed for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.
  • Deuterium labeled compound
  • Long-acting Sulfonamide
  • Used for coccidiosis treatment
  • Used for bacterial infections in poultry, swine, and cattle
  • Store as powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL