Learn More
Medchemexpress LLC Sulfabrom-d4 | 2374858-36-1 | 361.25 | 100 MG
SDP
Catalog No. 5000448438
Encompass_Preferred
Sulfabrom-d4 (N 3517-d4) is the deuterium labeled Sulfabrom (HY-U00131). Sulfabrom is a long-acting Sulfonamide used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.
  • Deuterium labeled Sulfabrom
  • Long-acting Sulfonamide
  • Used for coccidiosis treatment
  • Used for various bacterial infections in poultry, swine, and cattle
Catalog No. 50-004-48438 Supplier Medchemexpress LLC Supplier No. HYU00131S100MG
Sulfabrom-d4 (N 3517-d4) is the deuterium labeled Sulfabrom (HY-U00131). Sulfabrom is a long-acting Sulfonamide used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.
  • Deuterium labeled Sulfabrom
  • Long-acting Sulfonamide
  • Used for coccidiosis treatment
  • Used for various bacterial infections in poultry, swine, and cattle