Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfabrom-d4 | 2374858-36-1 | 361.25 | 5 MG
SDP

Catalog No. 5000448300
Encompass_Preferred

Sulfabrom-d4 (N 3517-d4) is the deuterium labeled Sulfabrom. It is a long-acting Sulfonamide used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.

  • Target: Isotope-labeled compounds
  • Pathway: Others
  • Solvent and solubility: DMSO at 100 mg/mL (276.82 mM); need ultrasonic
  • Caution: Product has not been fully validated for medical applications. For research use only.

Catalog No. 50-004-48300 Supplier Medchemexpress LLC Supplier No. HYU00131S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfabrom-d4 (N 3517-d4) is the deuterium labeled Sulfabrom. It is a long-acting Sulfonamide used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.

  • Target: Isotope-labeled compounds
  • Pathway: Others
  • Solvent and solubility: DMSO at 100 mg/mL (276.82 mM); need ultrasonic
  • Caution: Product has not been fully validated for medical applications. For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.