Learn More
Selleck Chemical LLC Sulfacetamide sodium salt hydrate S4750-100mg
SDP
Catalog No. 503111578
Encompass_Preferred
Sulfacetamide is a sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase(DHPS) with IC50 of 9 5 M Sulfacetamide is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA) which is required for bacterial synthesis of folic acid
Catalog No. 50-311-1578 Supplier Selleck Chemical LLC Supplier No. S4750100MG
Sulfacetamide is a sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase(DHPS) with IC50 of 9 5 M Sulfacetamide is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA) which is required for bacterial synthesis of folic acid