Learn More
Apexbio Technology LLC Sulfacetamide Sodium 127-56-0 5g
SDP
Catalog No. 5000091222
Encompass_Preferred
Sulfacetamide Sodium (127-56-0) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby disrupting bacterial folic acid synthesis Sulfacetamide Sodium exerts its biological activity primarily through inhibition of bacterial dihydropteroate synthase In in vitro studies Sulfacetamide Sodium demonstrates bacteriostatic activity with inhibitory concentrations (IC50) ranging from 60 g/mL to 64 mg/mL dependent on the bacterial species studied Based on these pharmacological properties Sulfacetamide Sodium holds research potential in microbiological studies of antibacterial potency antibiotic susceptibility and resistance patterns
Catalog No. 50-000-91222 Supplier Apexbio Technology LLC Supplier No. B13955000
Sulfacetamide Sodium (127-56-0) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby disrupting bacterial folic acid synthesis Sulfacetamide Sodium exerts its biological activity primarily through inhibition of bacterial dihydropteroate synthase In in vitro studies Sulfacetamide Sodium demonstrates bacteriostatic activity with inhibitory concentrations (IC50) ranging from 60 g/mL to 64 mg/mL dependent on the bacterial species studied Based on these pharmacological properties Sulfacetamide Sodium holds research potential in microbiological studies of antibacterial potency antibiotic susceptibility and resistance patterns