Learn More
Medchemexpress LLC Sulfacetamide sodium | 127-56-0 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB057610MM/1ML
Encompass_Preferred
Sulfacetamide sodium is a sulfonamide antibiotic used for the study of ocular infections. It exhibits antifungal and antibacterial activities. It can be used for the study of various ocular infections like trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea, acting as an antifungal agent through a CYP51A1-independent mechanism and as a bacteriostatic agent with similar activity against both Gram-positive and Gram-negative bacteria.
  • Sulfonamide antibiotic
  • Antifungal and antibacterial activities
  • Effective against various ocular infections including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
  • Antifungal action through a CYP51A1-independent mechanism
  • Bacteriostatic agent against Gram-positive and Gram-negative bacteria
Catalog No. 50-004-09025