Learn More
Medchemexpress LLC Sulfacetamide sodium monohydrate | 6209-17-2 | 99.5% | 25 G
SDP
Catalog No. 5000406576
Encompass_Preferred
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic used for the study of ocular infections. It exhibits antifungal and antibacterial activities, acting as a bacteriostatic agent against both Gram-positive and Gram-negative bacteria. This compound is suitable for researching various ocular infections such as trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea, and it functions as an antifungal drug via a CYP51A1-independent mechanism.
  • Exhibits antifungal activity
  • Exhibits antibacterial activity
  • Acts as a bacteriostatic agent
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Used for studying ocular infections including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
  • Functions as an antifungal drug via a CYP51A1-independent mechanism
  • For research use only
Catalog No. 50-004-06576 Supplier Medchemexpress LLC Supplier No. 25GHYB0888
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic used for the study of ocular infections. It exhibits antifungal and antibacterial activities, acting as a bacteriostatic agent against both Gram-positive and Gram-negative bacteria. This compound is suitable for researching various ocular infections such as trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea, and it functions as an antifungal drug via a CYP51A1-independent mechanism.
  • Exhibits antifungal activity
  • Exhibits antibacterial activity
  • Acts as a bacteriostatic agent
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Used for studying ocular infections including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
  • Functions as an antifungal drug via a CYP51A1-independent mechanism
  • For research use only