Learn More
Medchemexpress LLC Sulfacetamide sodium monohydrate | 6209-17-2 | 99.8% | 100 G
SDP
Catalog No. 5000409229
Encompass_Preferred
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic primarily used for research purposes. It demonstrates both antibacterial and antifungal activities, making it suitable for the study of various ocular infections.
  • Can be used for the study of various ocular infections, including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea.
  • Exhibits antifungal activity.
  • Acts as a bacteriostatic agent against Gram-positive and Gram-negative bacteria.
  • For research use only.
Catalog No. 50-004-09229 Supplier Medchemexpress LLC Supplier No. HYB0888100G
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic primarily used for research purposes. It demonstrates both antibacterial and antifungal activities, making it suitable for the study of various ocular infections.
  • Can be used for the study of various ocular infections, including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea.
  • Exhibits antifungal activity.
  • Acts as a bacteriostatic agent against Gram-positive and Gram-negative bacteria.
  • For research use only.