Learn More
Medchemexpress LLC Sulfacetamide sodium monohydrate | 6209-17-2 | 99.8% | 5 G
SDP
Catalog No. 5000408989
Encompass_Preferred
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic used for the study of ocular infections. It exhibits both antifungal and antibacterial activities, acting as a bacteriostatic agent against Gram-positive and Gram-negative bacteria. This compound is suitable for researching various ocular infections, including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea.
  • Sulfonamide antibiotic
  • Used for the study of ocular infections
  • Exhibits antifungal activity
  • Exhibits antibacterial activity
  • Bacteriostatic agent
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Acts through a CYP51A1-independent mechanism for antifungal action
  • Suitable for researching trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
Catalog No. 50-004-08989 Supplier Medchemexpress LLC Supplier No. HYB08885G
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic used for the study of ocular infections. It exhibits both antifungal and antibacterial activities, acting as a bacteriostatic agent against Gram-positive and Gram-negative bacteria. This compound is suitable for researching various ocular infections, including trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea.
  • Sulfonamide antibiotic
  • Used for the study of ocular infections
  • Exhibits antifungal activity
  • Exhibits antibacterial activity
  • Bacteriostatic agent
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
  • Acts through a CYP51A1-independent mechanism for antifungal action
  • Suitable for researching trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea