Learn More
Medchemexpress LLC Sulfacetamide sodium monohydrate | 6209-17-2 | MFCD00149555 | 99.8% | 50 G
SDP
Catalog No. 5000409218
Encompass_Preferred
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic exhibiting both antifungal and antibacterial activities. It acts as a bacteriostatic agent, effective against both Gram-positive and Gram-negative bacteria. This compound is utilized in the study of various ocular infections and related conditions.
  • Sulfonamide antibiotic
  • Exhibits antifungal and antibacterial activities
  • Bacteriostatic agent effective against Gram-positive and Gram-negative bacteria
  • Used in studies of ocular infections such as trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
  • Antifungal action through a CYP51A1-independent mechanism
  • For research use only
Catalog No. 50-004-09218 Supplier Medchemexpress LLC Supplier No. HYB088850G
Sulfacetamide sodium monohydrate is a sulfonamide antibiotic exhibiting both antifungal and antibacterial activities. It acts as a bacteriostatic agent, effective against both Gram-positive and Gram-negative bacteria. This compound is utilized in the study of various ocular infections and related conditions.
  • Sulfonamide antibiotic
  • Exhibits antifungal and antibacterial activities
  • Bacteriostatic agent effective against Gram-positive and Gram-negative bacteria
  • Used in studies of ocular infections such as trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor, and rosacea
  • Antifungal action through a CYP51A1-independent mechanism
  • For research use only