Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfacetamide sodium salt hydrate, 99%

Catalog No. AAB2242614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2242614 25 g
AAB2242636 500 g
AAB2242622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2242614 Supplier Thermo Scientific Chemicals Supplier No. B2242614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6209-17-2 | C8H9N2NaO3S | 236.22 g/mol

Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

Solubility
Freely soluble in water. Insoluble in chloroform, ether, and benzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6209-17-2
Molecular Formula C8H9N2NaO3S
Molecular Weight (g/mol) 236.22
MDL Number MFCD00007885
InChI Key PQMSFAORUFMASU-UHFFFAOYSA-M
Synonym sodium sulfacetamide, sulfacetamide sodium monohydrate, sulfacetamide sodium salt hydrate, sulfacetamide sodic hydrate, unii-4nrt660kjq, sulfacetamide sodium usp, cetamide, sulfair forte, sulfair 10, 4nrt660kjq
PubChem CID 6419954
ChEBI CHEBI:63858
IUPAC Name sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
SMILES CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Quantity 25 g
Solubility Information Freely soluble in water. Insoluble in chloroform,ether,and benzene.
Formula Weight 236.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sulfacetamide sodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-848-4
RTECSNumber AC8480000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.