Learn More
Medchemexpress LLC HY-B1781 250mg Medchemexpress, Sulfachloropyridazine CAS:80-32-0 Purity:>98%
SDP
Catalog No. 501933008
Encompass_Preferred
Medchemexpress, HY-B1781 250mg Sulfachloropyridazine CAS:80-32-0 Sulfachloropyridazine is a broad spectrum sulfonamide used against both Gram-positive and Gram-negative aerobic bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3008 Supplier Medchemexpress LLC Supplier No. HYB1781250MG
Medchemexpress, HY-B1781 250mg Sulfachloropyridazine CAS:80-32-0 Sulfachloropyridazine is a broad spectrum sulfonamide used against both Gram-positive and Gram-negative aerobic bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.