Learn More
Medchemexpress LLC Sulfachloropyridazine | 80-32-0 | 98.3% | 250 MG
SDP
Catalog No. 5000413954
Encompass_Preferred
Sulfachloropyridazine (Standard) is the analytical standard of Sulfachloropyridazine. It is a broad-spectrum sulfonamide effective against gram-positive and gram-negative aerobic bacteria and Chlamydia. It is also considered a common pollutant in surface and groundwater and is used as a model for sulfonamide antibiotics in electro-Fenton processes.
  • Intended for research and analytical applications
  • Broad-spectrum sulfonamide
  • Effective against gram-positive and gram-negative aerobic bacteria and Chlamydia
  • Used as a model for sulfonamide antibiotics in electro-Fenton processes
Catalog No. 50-004-13954 Supplier Medchemexpress LLC Supplier No. 250MGHYB1781R
Add to Cart
Add to Cart
Sulfachloropyridazine (Standard) is the analytical standard of Sulfachloropyridazine. It is a broad-spectrum sulfonamide effective against gram-positive and gram-negative aerobic bacteria and Chlamydia. It is also considered a common pollutant in surface and groundwater and is used as a model for sulfonamide antibiotics in electro-Fenton processes.
  • Intended for research and analytical applications
  • Broad-spectrum sulfonamide
  • Effective against gram-positive and gram-negative aerobic bacteria and Chlamydia
  • Used as a model for sulfonamide antibiotics in electro-Fenton processes