Learn More
Medchemexpress LLC HY-19285A 100mg Medchemexpress, Sulfaclozine (sodium) CAS:23307-72-4 Purity:>98%
SDP
Catalog No. 501970020
Encompass_Preferred
Medchemexpress, HY-19285A 100mg Sulfaclozine (sodium) CAS:23307-72-4 Sulfaclozine sodium (Sulfachloropyrazine sodium) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. Sulfaclozine sodium is commonly used for the treatment of various poultry diseases (particularly, collibacteriosis, fowl cholera and coccidiosis). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0020 Supplier Medchemexpress LLC Supplier No. HY19285A100MG
Medchemexpress, HY-19285A 100mg Sulfaclozine (sodium) CAS:23307-72-4 Sulfaclozine sodium (Sulfachloropyrazine sodium) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. Sulfaclozine sodium is commonly used for the treatment of various poultry diseases (particularly, collibacteriosis, fowl cholera and coccidiosis). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.