Learn More
Medchemexpress LLC Sulfaclozine sodium (sulfachloropyrazine sodium) | 23307-72-4 | 99.02% | 306.70 | 1 ML
SDP
Catalog No. 5000385562
Encompass_Preferred
Sulfaclozine sodium (Sulfachloropyrazine sodium) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. It is commonly used for the treatment of various poultry diseases, particularly collibacteriosis, fowl cholera, and coccidiosis.
  • Efficacious sulphonamide derivative
  • Antibacterial and anticoccidial effects
  • Used for treatment of various poultry diseases (collibacteriosis, fowl cholera, coccidiosis)
Catalog No. 50-003-85562 Supplier Medchemexpress LLC Supplier No. HY19285A10MM/1ML
Sulfaclozine sodium (Sulfachloropyrazine sodium) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. It is commonly used for the treatment of various poultry diseases, particularly collibacteriosis, fowl cholera, and coccidiosis.
  • Efficacious sulphonamide derivative
  • Antibacterial and anticoccidial effects
  • Used for treatment of various poultry diseases (collibacteriosis, fowl cholera, coccidiosis)