Learn More
Cayman Chemical Sulfaclozine (sodium salt), Purity: ≥98%, CAS: 23307-72-4, 500 mg
SDP
Catalog No. 502991469
Encompass_Preferred
A sulfonamide antibiotic with antiprotozoal activity; dietary administration reduces cecal lesion score and oocyst production in a chick model of E. tenella infection; increases average survival time in a mouse model of T. gondii infection at 250 mg/kg per day; has been found in chicken muscle samples from domestic markets and as an environmental contaminent in fish pond water, biogas digester water, and lagoon wastewater from pig farms
Catalog No. 50-299-1469 Supplier Cayman Chemical Supplier No. 31223500 MG
A sulfonamide antibiotic with antiprotozoal activity; dietary administration reduces cecal lesion score and oocyst production in a chick model of E. tenella infection; increases average survival time in a mouse model of T. gondii infection at 250 mg/kg per day; has been found in chicken muscle samples from domestic markets and as an environmental contaminent in fish pond water, biogas digester water, and lagoon wastewater from pig farms