Learn More
Cayman Chemical Sulfadiazine, Purity: ≥98%, CAS: 68-35-9, 100 g
SDP
Catalog No. 502995195
Encompass_Preferred
A sulfonamide antibiotic; inhibits recombinant P. carinii DHPS (IC50 = 0.19 μM); active against clinical isolates of M. tuberculosis (MIC90 = 10 mg/L) and of N. meningitidis (MICs = 5-2,000 mg/L); active against A. pleuropneumoniae, S. choleraesuis, S. typhimurium, P. multocida, S. equi, and S. suis (MIC90s = T. gondii infection when administered in combination with pyrimethamine at 40 mg/kg per day
Catalog No. 50-299-5195 Supplier Cayman Chemical Supplier No. 23719100 G
A sulfonamide antibiotic; inhibits recombinant P. carinii DHPS (IC50 = 0.19 μM); active against clinical isolates of M. tuberculosis (MIC90 = 10 mg/L) and of N. meningitidis (MICs = 5-2,000 mg/L); active against A. pleuropneumoniae, S. choleraesuis, S. typhimurium, P. multocida, S. equi, and S. suis (MIC90s = T. gondii infection when administered in combination with pyrimethamine at 40 mg/kg per day