Learn More
Cayman Chemical Sulfadiazine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1020719-78-1, 1 mg
SDP
Catalog No. 502946137
Encompass_Preferred
An internal standard for the quantification of sulfadiazine by GC- or LC-MS
Catalog No. 50-294-6137 Supplier Cayman Chemical Supplier No. 336051 MG
An internal standard for the quantification of sulfadiazine by GC- or LC-MS