Learn More
Cayman Chemical SulfadIazIn-d4 5mg
SDP
Supplier:  Cayman Chemical 336055 MG
Encompass_Preferred
An internal standard for the quantification of sulfadiazine by GC- or LC-MS
Catalog No. 50-294-6256