Learn More
Medchemexpress LLC Sulfadiazine-13C6 | 1189426-16-1 | 1 MG
SDP
Catalog No. 5000408999
Encompass_Preferred
Sulfadiazine-13C6 is a labeled Sulfadiazine, a sulfonamide antibiotic with antimalarial activity. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08999 Supplier Medchemexpress LLC Supplier No. HYB0273S11MG
Sulfadiazine-13C6 is a labeled Sulfadiazine, a sulfonamide antibiotic with antimalarial activity. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS