Learn More
Apexbio Technology LLC Sulfadiazine 68-35-9 5g
SDP
Catalog No. 5000093847
Encompass_Preferred
Sulfadiazine (CAS 68-35-9) is a small-molecule inhibitor targeting dihydropteroate synthase It is designed to inhibit microbial folate biosynthesis thereby disrupting folic acid synthesis required for pathogen proliferation Sulfadiazine exerts its biological activity primarily through the competitive inhibition of dihydropteroate synthase In in vitro studies Sulfadiazine demonstrates antiparasitic activity with IC50 values ranging from approximately 2 g/mL to 5 g/mL against Toxoplasma gondii depending on experimental conditions and parasite strains Based on these pharmacological properties Sulfadiazine holds research potential in antiparasitic activity evaluation and studies of folic acid metabolism pathways
Catalog No. 50-000-93847 Supplier Apexbio Technology LLC Supplier No. A85275000
Sulfadiazine (CAS 68-35-9) is a small-molecule inhibitor targeting dihydropteroate synthase It is designed to inhibit microbial folate biosynthesis thereby disrupting folic acid synthesis required for pathogen proliferation Sulfadiazine exerts its biological activity primarily through the competitive inhibition of dihydropteroate synthase In in vitro studies Sulfadiazine demonstrates antiparasitic activity with IC50 values ranging from approximately 2 g/mL to 5 g/mL against Toxoplasma gondii depending on experimental conditions and parasite strains Based on these pharmacological properties Sulfadiazine holds research potential in antiparasitic activity evaluation and studies of folic acid metabolism pathways