Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfadiazine | 68-35-9 | 99.9% | 1 G
SDP

Catalog No. 5000406992
Encompass_Preferred

Sulfadiazine is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research. With a molecular formula of C10H10N4O2S and a molecular weight of 250.28, this compound targets bacterial and parasitic organisms, functioning within the anti-infection pathway. It is available in solid form.

  • Purity is 99.89%.
  • Powder is stable for 3 years at -20°C and 2 years at 4°C.
  • In solvent, stable for 2 years at -80°C and 1 year at -20°C.
  • In vitro solubility: soluble in DMSO (≥ 50 mg/mL) but insoluble in H₂O.
  • In vivo solubility options include 10% DMSO with various diluents.
  • IC₅₀ and target information for Toxoplasma is available.

Catalog No. 50-004-06992 Supplier Medchemexpress LLC Supplier No. HYB02731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfadiazine is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research. With a molecular formula of C10H10N4O2S and a molecular weight of 250.28, this compound targets bacterial and parasitic organisms, functioning within the anti-infection pathway. It is available in solid form.

  • Purity is 99.89%.
  • Powder is stable for 3 years at -20°C and 2 years at 4°C.
  • In solvent, stable for 2 years at -80°C and 1 year at -20°C.
  • In vitro solubility: soluble in DMSO (≥ 50 mg/mL) but insoluble in H₂O.
  • In vivo solubility options include 10% DMSO with various diluents.
  • IC₅₀ and target information for Toxoplasma is available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.