Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfadiazine | 68-35-9 | 99.9% | 10 MM * 1 ML
SDP

Supplier:  Medchemexpress LLC HYB027310MM/1ML

Encompass_Preferred

Sulfadiazine is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research. This product is intended for research use only and has not been fully validated for medical applications.

  • Functions as a sulfonamide antibiotic
  • Exhibits antimalarial activity
  • Suitable for toxoplasmosis research
  • Supplied as a white to off-white solid
  • High purity of 99.9%

Catalog No. 50-004-08711


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.