Learn More
Medchemexpress LLC Sulfadiazine | 68-35-9 | 99.9%
SDP
Catalog No. 5000405578
Encompass_Preferred
Sulfadiazine is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research.
  • Functions as a sulfonamide antibiotic
  • Exhibits antimalarial properties
  • Utilized in toxoplasmosis research
  • Demonstrates activity against Toxoplasma gondii in various cell lines (e.g., MRC5, Vero)
  • Effectiveness evaluated in in vivo models against T. gondii strains
Catalog No. 50-004-05578 Supplier Medchemexpress LLC Supplier No. 250MGHYB0273R
Sulfadiazine is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research.
  • Functions as a sulfonamide antibiotic
  • Exhibits antimalarial properties
  • Utilized in toxoplasmosis research
  • Demonstrates activity against Toxoplasma gondii in various cell lines (e.g., MRC5, Vero)
  • Effectiveness evaluated in in vivo models against T. gondii strains