Learn More
Sulfadiazine, 98%
Description
Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)
Solubility
Soluble in dimethyl sulfoxide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 253°C to 256°C |
| Assay Percent Range | 99% |
| Quantity | 1000 g |
| Merck Index | 14,8903 |
| Solubility Information | Soluble in dimethyl sulfoxide. |
| Formula Weight | 250.28 |
| Percent Purity | 99% |
| Chemical Name or Material | Sulfadiazine |
Safety and Handling
| missing translation for 'healthHazard1' | H302-H315-H317-H319-H334-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H334-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c |
| EINECSNumber | 200-685-8 |
| RTECSNumber | WP1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.