Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfadiazine sodium | 547-32-0 | >98.0% | 1 ML
SDP

Catalog No. 5000408974
Encompass_Preferred

Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research.

  • Antimicrobial properties for various infections
  • Effective in toxoplasmosis research
  • Purity >98.0%
  • White to off-white solid appearance
  • Molecular weight of 272.26
  • Chemical formula C10H9N4NaO2S

Catalog No. 50-004-08974 Supplier Medchemexpress LLC Supplier No. HYB0273A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfadiazine sodium is a sulfonamide antibiotic with antimalarial activity, primarily used for toxoplasmosis research.

  • Antimicrobial properties for various infections
  • Effective in toxoplasmosis research
  • Purity >98.0%
  • White to off-white solid appearance
  • Molecular weight of 272.26
  • Chemical formula C10H9N4NaO2S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.